Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adig
Entrez Gene ID
100765200
Gene Name
adipogenin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613641: 2362175 - 2369897 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007607969.1 XP_007606159.1 3 - 4 286 95

View Full Product Details