Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Podxl2
Entrez Gene ID
100768042
Gene Name
podocalyxin-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613782: 302974 - 326416 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640959.1 XP_007639149.1 2 - 3 1035 68

View Full Product Details