Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chst7
Entrez Gene ID
103163077
Gene Name
carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 7
Gene Aliases
I79_011958
Chromosome Location
Chr.Un|NW_003614094: 233423 - 234768 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645343.1 XP_007643533.1 1 - 2 195 110

View Full Product Details