Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prf1
Entrez Gene ID
100761729
Gene Name
perforin 1 (pore forming protein)
Gene Aliases
Perforin-1
Chromosome Location
Chr.Un|NW_003613759: 696857 - 700338 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499881.1 XP_003499929.1 1 - 2 539 85

View Full Product Details