Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cacng6
Entrez Gene ID
100763524
Gene Name
calcium channel, voltage-dependent, gamma subunit 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614747: 223696 - 256436 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650268.1 XP_007648458.1 1 - 2 96 64

View Full Product Details