Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss54
Entrez Gene ID
100760622
Gene Name
protease, serine, 54
Gene Aliases
I79_017770
Chromosome Location
Chr.Un|NW_003614730: 179187 - 215386 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509573.2 XP_003509621.1 5 - 6 863 89
XM_007650190.1 XP_007648380.1 5 - 6 1078 89
XM_007650191.1 XP_007648381.1 4 - 5 691 89

View Full Product Details