Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyg1
Entrez Gene ID
100762750
Gene Name
lysozyme G-like 1
Gene Aliases
I79_019279
Chromosome Location
Chr.Un|NW_003614980: 12280 - 19274 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510864.1 XP_003510912.1 1 - 2 40 66

View Full Product Details