Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppapdc2
Entrez Gene ID
103162885
Gene Name
phosphatidic acid phosphatase type 2 domain containing 2
Gene Aliases
I79_013643
Chromosome Location
Chr.Un|NW_003614228: 801663 - 802720 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646816.1 XP_007645006.1 1 - 1 422 69

View Full Product Details