Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tlr5
Entrez Gene ID
100772103
Gene Name
toll-like receptor 5
Gene Aliases
I79_000060
Chromosome Location
Chr.Un|NW_003613581: 1231068 - 1234341 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003494963.1 XP_003495011.1 1 - 2 36 62

View Full Product Details