Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smtnl1
Entrez Gene ID
100757041
Gene Name
smoothelin-like 1
Gene Aliases
I79_013454
Chromosome Location
Chr.Un|NW_003614216: 527478 - 535217 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506157.1 XP_003506205.1 1 - 2 633 70

View Full Product Details