Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkrip1
Entrez Gene ID
100766978
Gene Name
PRKR interacting protein 1 (IL11 inducible)
Gene Aliases
I79_007280
Chromosome Location
Chr.Un|NW_003613822: 1046001 - 1077075 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500956.2 XP_003501004.1 2 - 3 97 81

View Full Product Details