Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Awat2
Entrez Gene ID
100768365
Gene Name
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases
I79_018025
Chromosome Location
Chr.Un|NW_003614772: 448453 - 459864 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509812.1 XP_003509860.1 1 - 2 85 66

View Full Product Details