Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl11b
Entrez Gene ID
100752444
Gene Name
methyltransferase like 11B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614159: 961596 - 978837 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505528.2 XP_003505576.1 3 - 4 572 88

View Full Product Details