Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brs3
Entrez Gene ID
100763382
Gene Name
bombesin-like receptor 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613755: 1305283 - 1309502 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499816.1 XP_003499864.1 2 - 3 788 87

View Full Product Details