Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tp53inp2
Entrez Gene ID
100751556
Gene Name
tumor protein p53 inducible nuclear protein 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613833: 1053525 - 1056485 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501127.2 XP_003501175.1 1 - 2 755 69

View Full Product Details