Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb47
Entrez Gene ID
100767262
Gene Name
zinc finger and BTB domain containing 47
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613803: 1115585 - 1123906 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641432.1 XP_007639622.1 1 - 2 320 100

View Full Product Details