Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lancl3
Entrez Gene ID
100762249
Gene Name
LanC lantibiotic synthetase component C-like 3 (bacterial)
Gene Aliases
I79_016004
Chromosome Location
Chr.Un|NW_003614493: 262056 - 367860 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508157.1 XP_003508205.1 3 - 4 302 107

View Full Product Details