Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam29
Entrez Gene ID
100759070
Gene Name
ADAM metallopeptidase domain 29
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614571: 275608 - 277886 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649307.1 XP_007647497.1 1 - 1 2222 80

View Full Product Details