Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Htr2b
Entrez Gene ID
103161688
Gene Name
5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled
Gene Aliases
I79_024744
Chromosome Location
Chr.Un|NW_003617646: 41327 - 53218 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655248.1 XP_007653438.1 1 - 2 332 77

View Full Product Details