Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrfap1
Entrez Gene ID
100769822
Gene Name
Morf4 family associated protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613629: 2036561 - 2038467 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496616.2 XP_003496664.1 1 - 2 545 62

View Full Product Details