Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Panx3
Entrez Gene ID
100754757
Gene Name
pannexin 3
Gene Aliases
Pannexin-3
Chromosome Location
Chr.Un|NW_003616556: 105727 - 112886 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514369.2 XP_003514417.1 3 - 4 542 78

View Full Product Details