Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk13
Entrez Gene ID
100761429
Gene Name
kallikrein-related peptidase 13
Gene Aliases
I79_025194
Chromosome Location
Chr.Un|NW_003618538: 19016 - 25240 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007607730.1 XP_007605920.1 1 - 2 122 76

View Full Product Details