Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nynrin
Entrez Gene ID
100768139
Gene Name
NYN domain and retroviral integrase containing
Gene Aliases
I79_008525
Chromosome Location
Chr.Un|NW_003613875: 863447 - 879152 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502082.1 XP_003502130.1 2 - 3 850 62

View Full Product Details