Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl6ip6
Entrez Gene ID
100774425
Gene Name
ADP-ribosylation-like factor 6 interacting protein 6
Gene Aliases
I79_021854
Chromosome Location
Chr.Un|NW_003615674: 213020 - 218346 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512963.2 XP_003513011.2 2 - 3 476 127

View Full Product Details