Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heatr4
Entrez Gene ID
100752922
Gene Name
HEAT repeat containing 4
Gene Aliases
I79_012297
Chromosome Location
Chr.Un|NW_003614123: 486215 - 520781 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505142.2 XP_003505190.1 10 - 11 3141 71
XM_007645667.1 XP_007643857.1 11 - 12 3228 71
XM_007645668.1 XP_007643858.1 10 - 11 2260 71
XM_007645669.1 XP_007643859.1 11 - 12 2318 71
XM_007645670.1 XP_007643860.1 11 - 12 2284 71
XM_007645671.1 XP_007643861.1 11 - 12 2362 71
XM_007645672.1 XP_007643862.1 10 - 11 3066 71

View Full Product Details