Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heatr4
Entrez Gene ID
100752922
Gene Name
HEAT repeat containing 4
Gene Aliases
I79_012297
Chromosome Location
Chr.Un|NW_003614123: 486215 - 520781 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505142.2 XP_003505190.1 14 - 15 3669 106
XM_007645667.1 XP_007643857.1 15 - 16 3756 106
XM_007645668.1 XP_007643858.1 14 - 15 2788 106
XM_007645669.1 XP_007643859.1 15 - 16 2846 106
XM_007645670.1 XP_007643860.1 15 - 16 2812 106
XM_007645671.1 XP_007643861.1 15 - 16 2890 106
XM_007645672.1 XP_007643862.1 14 - 15 3594 106

View Full Product Details