Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Samd1
Entrez Gene ID
100760482
Gene Name
sterile alpha motif domain containing 1
Gene Aliases
Atherin, I79_008383
Chromosome Location
Chr.Un|NW_003613865: 1285522 - 1289087 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642567.1 XP_007640757.1 5 - 6 782 93
GenBank mRNA JI882104.1 - 3 - 4 237 93

View Full Product Details