Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr87
Entrez Gene ID
100762812
Gene Name
WD repeat domain 87
Gene Aliases
I79_010740
Chromosome Location
Chr.Un|NW_003614005: 89893 - 108851 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644397.1 XP_007642587.1 4 - 5 3420 64

View Full Product Details