Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd8b
Entrez Gene ID
100769972
Gene Name
CD8b molecule
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614012: 754034 - 764779 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503913.1 XP_003503961.1 2 - 3 404 73

View Full Product Details