Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spem1
Entrez Gene ID
100764619
Gene Name
spermatid maturation 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613643: 1880217 - 1881820 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497184.2 XP_003497232.1 2 - 3 604 76

View Full Product Details