Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Evx2
Entrez Gene ID
100759401
Gene Name
even-skipped homeobox 2
Gene Aliases
I79_004134
Chromosome Location
Chr.Un|NW_003613688: 1486015 - 1497234 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498340.1 XP_003498388.1 1 - 2 428 62

View Full Product Details