Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klrg2
Entrez Gene ID
100771105
Gene Name
killer cell lectin-like receptor subfamily G, member 2
Gene Aliases
I79_009966
Chromosome Location
Chr.Un|NW_003613951: 312552 - 325529 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643833.1 XP_007642023.1 2 - 3 220 91

View Full Product Details