Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nipal1
Entrez Gene ID
100774364
Gene Name
NIPA-like domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617853: 39125 - 50342 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515355.1 XP_003515403.1 1 - 2 265 65

View Full Product Details