Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uncx
Entrez Gene ID
100771344
Gene Name
UNC homeobox
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613618: 4207361 - 4213724 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496324.1 XP_003496372.1 1 - 2 84 115

View Full Product Details