Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kprp
Entrez Gene ID
100756266
Gene Name
keratinocyte proline-rich protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614331: 211777 - 213818 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507101.2 XP_003507149.1 1 - 1 1819 69

View Full Product Details