Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmrtb1
Entrez Gene ID
100772202
Gene Name
DMRT-like family B with proline-rich C-terminal, 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613599: 4323213 - 4330058 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495744.1 XP_003495792.1 3 - 4 732 86

View Full Product Details