Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2b2
Entrez Gene ID
100767263
Gene Name
SH2B adaptor protein 2
Gene Aliases
I79_007282
Chromosome Location
Chr.Un|NW_003613822: 1078112 - 1090597 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500957.1 XP_003501005.1 7 - 8 1020 108

View Full Product Details