Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd3eap
Entrez Gene ID
100755473
Gene Name
CD3e molecule, epsilon associated protein
Gene Aliases
I79_012655
Chromosome Location
Chr.Un|NW_003614150: 584180 - 592129 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645971.1 XP_007644161.1 2 - 3 105 80

View Full Product Details