Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnp1
Entrez Gene ID
100755165
Gene Name
transition protein 1 (during histone to protamine replacement)
Gene Aliases
I79_004991
Chromosome Location
Chr.Un|NW_003613713: 1574093 - 1574458 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498998.1 XP_003499046.1 1 - 2 140 61

View Full Product Details