Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Haus3
Entrez Gene ID
100770464
Gene Name
HAUS augmin-like complex, subunit 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614595: 592959 - 604904 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508850.2 XP_003508898.1 4 - 5 1820 119
XM_007649465.1 XP_007647655.1 4 - 5 2277 119
XM_007649466.1 XP_007647656.1 5 - 6 1874 119
GenBank mRNA JI870179.1 - 3 - 4 1239 119
JP043833.1 - 3 - 4 1239 119

View Full Product Details