Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gli4
Entrez Gene ID
100758024
Gene Name
GLI family zinc finger 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614880: 276249 - 281668 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510440.2 XP_003510488.1 3 - 4 273 78

View Full Product Details