Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl46
Entrez Gene ID
100770403
Gene Name
mitochondrial ribosomal protein L46
Gene Aliases
I79_023459
Chromosome Location
Chr.Un|NW_003616480: 4856 - 13265 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514274.2 XP_003514322.2 2 - 3 245 90

View Full Product Details