Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pspn
Entrez Gene ID
100761935
Gene Name
persephin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613861: 1187471 - 1188034 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501767.1 XP_003501815.1 1 - 2 159 97

View Full Product Details