Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcng4
Entrez Gene ID
100773553
Gene Name
potassium voltage-gated channel, subfamily G, member 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613583: 3918700 - 3928331 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495117.1 XP_003495165.1 1 - 2 748 73

View Full Product Details