Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lpar2
Entrez Gene ID
100753903
Gene Name
lysophosphatidic acid receptor 2
Gene Aliases
I79_005062
Chromosome Location
Chr.Un|NW_003613716: 141955 - 146486 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499062.2 XP_003499110.1 2 - 3 894 115

View Full Product Details