Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb8
Entrez Gene ID
100753972
Gene Name
ankyrin repeat and SOCS box containing 8
Gene Aliases
I79_021780
Chromosome Location
Chr.Un|NW_003615647: 34724 - 42243 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512902.2 XP_003512950.1 2 - 3 224 107
XM_007653293.1 XP_007651483.1 2 - 3 228 107
XM_007653294.1 XP_007651484.1 2 - 3 269 107

View Full Product Details