Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlx1
Entrez Gene ID
100768966
Gene Name
distal-less homeobox 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613606: 2737558 - 2741924 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495957.2 XP_003496005.1 2 - 3 842 62

View Full Product Details