Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp41
Entrez Gene ID
100757735
Gene Name
ZFP41 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614880: 291172 - 302059 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651078.1 XP_007649268.1 2 - 3 2206 108
XM_007651080.1 XP_007649270.1 2 - 3 2533 108
XM_007651081.1 XP_007649271.1 2 - 3 2144 108
XM_007651083.1 XP_007649273.1 2 - 3 2206 108
XM_007651084.1 XP_007649274.1 2 - 3 2206 108
XM_007651085.1 XP_007649275.1 2 - 3 2206 108

View Full Product Details