Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Svip
Entrez Gene ID
100765826
Gene Name
small VCP/p97-interacting protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614113: 256153 - 264443 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505033.2 XP_003505081.1 2 - 3 111 92

View Full Product Details