Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1qtnf2
Entrez Gene ID
100765047
Gene Name
C1q and tumor necrosis factor related protein 2
Gene Aliases
I79_008746
Chromosome Location
Chr.Un|NW_003613882: 917366 - 933953 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502208.2 XP_003502256.1 3 - 4 389 91

View Full Product Details